Assay Details
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284254.2 | XP_015139740.1 | 4 - 5 | 578 | 77 | |
XM_015284255.2 | XP_015139741.1 | 2 - 3 | 309 | 77 | ||
XM_015284256.2 | XP_015139742.1 | 3 - 4 | 399 | 77 | ||
XM_419684.6 | XP_419684.3 | 4 - 5 | 341 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
