Assay Details
| Gene Symbol: RGPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154658.1 | XP_025010426.1 | 2 - 3 | 1129 | 113 | |
| XM_025154659.1 | XP_025010427.1 | 2 - 3 | 1129 | 113 | ||
| Gene Symbol: LOC112533198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154084.1 | XP_025009852.1 | 3 - 4 | 113 | ||
More Information
Set Membership: |
|

RGPD1
Genomic Map