Assay Details
| Gene Symbol: FUT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144347.1 | XP_025000115.1 | 1 - 2 | 204 | 158 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025144347.1 transcripts.
Set Membership: |
|

FUT6
Genomic Map