Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232566.5 | XP_001232567.2 | 2 - 3 | 250 | 54 | |
| XM_004949751.3 | XP_004949808.1 | 2 - 3 | 817 | 54 | ||
| XM_015300404.2 | XP_015155890.1 | 2 - 3 | 329 | 54 | ||
| XM_015300405.2 | XP_015155891.1 | 2 - 3 | 258 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015300404.2 transcripts.
Set Membership: |
|

NEMP1
Genomic Map