Assay Details
Gene Symbol: C10orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025148376.1 | XP_025004144.1 | 1 - 2 | 10394 | 112 | |
XM_025148377.1 | XP_025004145.1 | 2 - 3 | 4655 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
