Assay Details
Gene Symbol: ST6GALNAC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025141711.1 | XP_024997479.1 | 3 - 4 | 732 | 60 | |
XM_420078.5 | XP_420078.3 | 3 - 4 | 667 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
