Assay Details
Gene Symbol: ST6GALNAC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946263.3 | XP_004946320.1 | 8 - 9 | 1317 | 91 | |
XM_025141711.1 | XP_024997479.1 | 8 - 9 | 1357 | 91 | ||
XM_420078.5 | XP_420078.3 | 8 - 9 | 1292 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
