Assay Details
| Gene Symbol: METTL21EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232406.4 | XP_001232407.2 | 4 - 5 | 554 | 139 | |
| XM_004938556.3 | XP_004938613.1 | 5 - 6 | 632 | 139 | ||
| XM_015275041.2 | XP_015130527.1 | 5 - 6 | 632 | 139 | ||
More Information
Set Membership: |
|

METTL21EP
Genomic Map