Assay Details
| Gene Symbol: QRFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235088.4 | XP_001235089.3 | 1 | 683 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

QRFP| Gene Symbol: QRFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235088.4 | XP_001235089.3 | 1 | 683 | 66 | |
Set Membership: |
|