Assay Details
Gene Symbol: CCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015273014.2 | XP_015128500.2 | 5 - 6 | 625 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
