Assay Details
| Gene Symbol: PGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642812.4 | XP_003642860.1 | 7 - 8 | 935 | 58 | |
| XM_025144061.1 | XP_024999829.1 | 7 - 8 | 978 | 58 | ||
More Information
Set Membership: |
|

PGAP3
Genomic Map