Assay Details
| Gene Symbol: IL20RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294596.2 | XP_015150082.1 | 5 - 6 | 850 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL20RB| Gene Symbol: IL20RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294596.2 | XP_015150082.1 | 5 - 6 | 850 | 60 | |
Set Membership: |
|