Assay Details
| Gene Symbol: LOC101751717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298210.2 | XP_015153696.1 | 1 - 2 | 1836 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101751717| Gene Symbol: LOC101751717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298210.2 | XP_015153696.1 | 1 - 2 | 1836 | 104 | |
Set Membership: |
|