Assay Details
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004934900.3 | XP_004934957.3 | 527 | 62 | ||
XM_025143916.1 | XP_024999684.1 | 2670 | 62 | |||
XM_025143917.1 | XP_024999685.1 | 2670 | 62 | |||
XM_025143918.1 | XP_024999686.1 | 2650 | 62 | |||
XM_025143919.1 | XP_024999687.1 | 2650 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
