Assay Details
| Gene Symbol: LOC771362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288926.2 | XP_015144412.1 | 12 - 13 | 1540 | 91 | |
| XM_015288927.2 | XP_015144413.1 | 12 - 13 | 1589 | 91 | ||
More Information
Set Membership: |
|

LOC771362
Genomic Map