Assay Details
| Gene Symbol: LOC771362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288926.2 | XP_015144412.1 | 6 - 7 | 773 | 100 | |
| XM_015288927.2 | XP_015144413.1 | 6 - 7 | 822 | 100 | ||
More Information
Set Membership: |
|

LOC771362
Genomic Map