Assay Details
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299123.2 | XP_015154609.1 | 1082 | 56 | ||
XM_015299124.2 | XP_015154610.1 | 1082 | 56 | |||
XM_015299125.2 | XP_015154611.1 | 1084 | 56 | |||
XM_015299126.2 | XP_015154612.1 | 1085 | 56 | |||
XM_015299127.2 | XP_015154613.1 | 1076 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
