Assay Details
| Gene Symbol: LOC100859726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300398.2 | XP_015155884.2 | 3 - 4 | 1006 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100859726| Gene Symbol: LOC100859726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300398.2 | XP_015155884.2 | 3 - 4 | 1006 | 53 | |
Set Membership: |
|