Assay Details
| Gene Symbol: BRICD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294468.2 | XP_015149954.1 | 3 - 4 | 1777 | 107 | |
| XM_025155247.1 | XP_025011015.1 | 5 - 6 | 878 | 107 | ||
More Information
Set Membership: |
|

BRICD5
Genomic Map