Assay Details
| Gene Symbol: BRICD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294467.2 | XP_015149953.1 | 4 - 5 | 1955 | 101 | |
| XM_015294468.2 | XP_015149954.1 | 4 - 5 | 1931 | 101 | ||
| XM_025155247.1 | XP_025011015.1 | 6 - 7 | 1032 | 101 | ||
More Information
Set Membership: |
|

BRICD5
Genomic Map