Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015300348.2 | XP_015155834.1 | 5 - 6 | 555 | 67 | |
XM_015300349.2 | XP_015155835.1 | 4 - 5 | 471 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
