Assay Details
Gene Symbol: LOC107049580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015273309.2 | XP_015128795.1 | 2 | 183 | 66 | |
XM_015273310.2 | XP_015128796.1 | 2 | 216 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
