Assay Details
Gene Symbol: SRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249715.1 | NP_001236644.1 | 1 - 2 | 357 | 67 | |
GenBank mRNA | AB000129.1 | - | 1 - 2 | 357 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
