Assay Details
Gene Symbol: SS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250596.1 | NP_001237525.1 | 11 - 12 | 2322 | 98 | |
GenBank mRNA | AF030231.1 | - | 11 - 12 | 2322 | 98 | |
AK285864.1 | - | 11 - 12 | 2333 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
