Assay Details
Gene Symbol: SS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250596.1 | NP_001237525.1 | 6 - 7 | 1075 | 91 | |
GenBank mRNA | AF030231.1 | - | 6 - 7 | 1075 | 91 | |
AK285864.1 | - | 6 - 7 | 1086 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
