Assay Details
| Gene Symbol: AAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250612.1 | NP_001237541.1 | 11 - 12 | 1437 | 98 | |
| GenBank mRNA | AF034210.1 | - | 11 - 12 | 1437 | 98 | |
| AK243764.1 | - | 11 - 12 | 1197 | 98 | ||
| AK244943.1 | - | 11 - 12 | 1206 | 98 | ||
More Information
Set Membership: |
|

AAT
Genomic Map