Assay Details
| Gene Symbol: AAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250612.1 | NP_001237541.1 | 2 - 3 | 434 | 66 | |
| GenBank mRNA | AF034210.1 | - | 2 - 3 | 434 | 66 | |
| AK243764.1 | - | 2 - 3 | 194 | 66 | ||
| AK244943.1 | - | 2 - 3 | 203 | 66 | ||
| Gene Symbol: LOC100804162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT097064.1 | - | 2 - 3 | 206 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT097064.1 transcripts.
Set Membership: |
|

AAT
Genomic Map