Assay Details
| Gene Symbol: AAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250612.1 | NP_001237541.1 | 8 - 9 | 1102 | 89 | |
| GenBank mRNA | AF034210.1 | - | 8 - 9 | 1102 | 89 | |
| AK243764.1 | - | 8 - 9 | 862 | 89 | ||
| AK244943.1 | - | 8 - 9 | 871 | 89 | ||
More Information
Set Membership: |
|

AAT
Genomic Map