Assay Details
| Gene Symbol: AAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250612.1 | NP_001237541.1 | 9 - 10 | 1224 | 64 | |
| GenBank mRNA | AF034210.1 | - | 9 - 10 | 1224 | 64 | |
| AK243764.1 | - | 9 - 10 | 984 | 64 | ||
| AK244943.1 | - | 9 - 10 | 993 | 64 | ||
More Information
Set Membership: |
|

AAT
Genomic Map