Assay Details
Gene Symbol: GLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF039851.1 | - | 2 - 3 | 140 | 79 | |
AK287031.1 | - | 11 - 12 | 1964 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
