Assay Details
Gene Symbol: GLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF039851.1 | - | 4 - 5 | 521 | 74 | |
AK287031.1 | - | 13 - 14 | 2345 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
