Assay Details
| Gene Symbol: PAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250853.1 | NP_001237782.1 | 6 - 7 | 1415 | 99 | |
| GenBank mRNA | AF089851.1 | - | 6 - 7 | 1415 | 99 | |
More Information
Set Membership: |
|

PAO1
Genomic Map