Assay Details
| Gene Symbol: PARP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250657.1 | NP_001237586.1 | 6 - 7 | 788 | 62 | |
| GenBank mRNA | AF169023.1 | - | 6 - 7 | 788 | 62 | |
More Information
Set Membership: |
|

PARP3
Genomic Map