Assay Details
| Gene Symbol: UBC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250674.1 | NP_001237603.1 | 3 - 4 | 258 | 126 | |
| GenBank mRNA | AF180143.1 | - | 3 - 4 | 258 | 126 | |
| Gene Symbol: LOC100804969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244076.1 | - | 3 - 4 | 406 | 126 | |
| AK245474.1 | - | 3 - 4 | 405 | 126 | ||
More Information
Set Membership: |
|

UBC4
Genomic Map