Assay Details
Gene Symbol: SCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250255.1 | NP_001237184.1 | 3 - 4 | 2457 | 71 | |
GenBank mRNA | AF195029.1 | - | 3 - 4 | 2457 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
