Assay Details
| Gene Symbol: CHIA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251632.1 | NP_001238561.1 | 2 - 3 | 614 | 61 | |
| GenBank mRNA | AF202731.1 | - | 2 - 3 | 614 | 61 | |
More Information
Set Membership: |
|

CHIA1
Genomic Map