Assay Details
Gene Symbol: LOC547605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248337.1 | NP_001235266.1 | 1 - 2 | 209 | 68 | |
GenBank mRNA | AF303940.1 | - | 1 - 2 | 209 | 68 | |
AF303941.1 | - | 1 - 2 | 209 | 68 | ||
AK243754.1 | - | 1 - 2 | 224 | 68 | ||
AK244118.1 | - | 1 - 2 | 220 | 68 | ||
AK285664.1 | - | 1 - 2 | 220 | 68 |
Gene Symbol: LOC100170694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK245366.1 | - | 1371 | 68 |
Gene Symbol: LOC100794277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254208.1 | NP_001241137.1 | 1 - 2 | 209 | 68 | |
GenBank mRNA | AF303939.1 | - | 1 - 2 | 209 | 68 | |
AK243761.1 | - | 1 - 2 | 224 | 68 | ||
AK244157.1 | - | 1 - 2 | 222 | 68 | ||
AK286277.1 | - | 1 - 2 | 224 | 68 | ||
U39567.1 | - | 1 - 2 | 206 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
