Assay Details
| Gene Symbol: LOC547605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248337.1 | NP_001235266.1 | 2 - 3 | 338 | 79 | |
| GenBank mRNA | AF303940.1 | - | 2 - 3 | 338 | 79 | |
| AF303941.1 | - | 2 - 3 | 338 | 79 | ||
| AK243754.1 | - | 2 - 3 | 353 | 79 | ||
| AK244118.1 | - | 2 - 3 | 349 | 79 | ||
| AK285664.1 | - | 2 - 3 | 349 | 79 | ||
| BT089205.1 | - | 2 - 3 | 354 | 79 | ||
| Gene Symbol: LOC100170694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK245366.1 | - | 1500 | 79 | ||
| Gene Symbol: LOC100794277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254208.1 | NP_001241137.1 | 2 - 3 | 338 | 79 | |
| GenBank mRNA | AF303939.1 | - | 2 - 3 | 338 | 79 | |
| AK243761.1 | - | 2 - 3 | 353 | 79 | ||
| AK244157.1 | - | 2 - 3 | 351 | 79 | ||
| AK286277.1 | - | 2 - 3 | 353 | 79 | ||
| U39567.1 | - | 2 - 3 | 335 | 79 | ||
More Information
Set Membership: |
|

LOC547605
Genomic Map