Assay Details
| Gene Symbol: CCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248512.2 | NP_001235441.1 | 4 - 5 | 503 | 73 | |
| GenBank mRNA | AF329816.1 | - | 4 - 5 | 432 | 73 | |
| FK017408.1 | - | 4 - 5 | 418 | 73 | ||
More Information
Set Membership: |
|

CCS
Genomic Map