Assay Details
| Gene Symbol: DR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248749.2 | NP_001235678.1 | 4 - 5 | 361 | 63 | |
| GenBank mRNA | AF464906.1 | - | 4 - 5 | 399 | 63 | |
| AK244706.1 | - | 4 - 5 | 402 | 63 | ||
| BT089816.1 | - | 4 - 5 | 363 | 63 | ||
More Information
Set Membership: |
|

DR1
Genomic Map