Assay Details
| Gene Symbol: DR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248749.2 | NP_001235678.1 | 5 - 6 | 433 | 89 | |
| GenBank mRNA | AF464906.1 | - | 5 - 6 | 471 | 89 | |
| AK244706.1 | - | 5 - 6 | 474 | 89 | ||
| BT089816.1 | - | 5 - 6 | 435 | 89 | ||
More Information
Set Membership: |
|

DR1
Genomic Map