Assay Details
| Gene Symbol: STP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251007.1 | NP_001237936.1 | 3 - 4 | 1153 | 72 | |
| GenBank mRNA | AJ563365.1 | - | 3 - 4 | 1153 | 72 | |
More Information
Set Membership: |
|

STP1
Genomic Map