Assay Details
| Gene Symbol: ODC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249411.1 | NP_001236340.1 | 1 | 1342 | 118 | |
| GenBank mRNA | AJ563383.1 | - | 1 | 1342 | 118 | |
More Information
Set Membership: |
|

ODC2
Genomic Map