Assay Details
Gene Symbol: PUR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251130.1 | NP_001238059.1 | 6 - 7 | 1206 | 80 | |
GenBank mRNA | AY321444.1 | - | 6 - 7 | 1206 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
