Assay Details
| Gene Symbol: GAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254052.1 | NP_001240981.1 | 5 - 6 | 966 | 73 | |
| GenBank mRNA | AB240966.1 | - | 5 - 6 | 966 | 73 | |
| AK285497.1 | - | 5 - 6 | 1027 | 73 | ||
| AY323131.1 | - | 4 - 5 | 808 | 73 | ||
More Information
Set Membership: |
|

GAD
Genomic Map