Assay Details
Gene Symbol: AGL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250104.1 | NP_001237033.1 | 1 - 2 | 451 | 110 | |
GenBank mRNA | AY370659.1 | - | 1 - 2 | 451 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
