Assay Details
Gene Symbol: AGL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250104.1 | NP_001237033.1 | 6 - 7 | 754 | 83 | |
GenBank mRNA | AY370659.1 | - | 6 - 7 | 754 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
