Assay Details
| Gene Symbol: CCS52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250239.1 | NP_001237168.1 | 7 - 8 | 1471 | 71 | |
| GenBank mRNA | AY439099.1 | - | 7 - 8 | 1471 | 71 | |
More Information
Set Membership: |
|

CCS52
Genomic Map