Assay Details
| Gene Symbol: FBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250386.1 | NP_001237315.1 | 2 - 3 | 703 | 107 | |
| GenBank mRNA | AK285598.1 | - | 2 - 3 | 755 | 107 | |
| AY492006.1 | - | 2 - 3 | 703 | 107 | ||
| Gene Symbol: LOC100792394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243904.1 | - | 1 - 2 | 1065 | 107 | |
| AK285169.1 | - | 2 - 3 | 755 | 107 | ||
More Information
Set Membership: |
|

FBA
Genomic Map