Assay Details
| Gene Symbol: LOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250756.1 | NP_001237685.1 | 1 - 2 | 295 | 80 | |
| GenBank mRNA | D13949.1 | - | 1 - 2 | 295 | 80 | |
| J03211.1 | - | 1 - 2 | 321 | 80 | ||
| JQ929619.1 | - | 1 - 2 | 240 | 80 | ||
More Information
Set Membership: |
|

LOX2
Genomic Map